Learn More
Electron Microscopy Sciences MeshWell Plate 24-well with Cluster
SDP
Catalog No. 5030869
Encompass_Preferred
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening. Includes one MeshWell and one Cluster plate.
Catalog No. 50-308-69 Supplier Electron Microscopy Sciences Supplier No. 6470224
Add to Cart
Add to Cart
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening. Includes one MeshWell and one Cluster plate.