Learn More
Biocare Medical, LLC MSH2 0.5ML
SDP
Supplier:  Biocare Medical, LLC CM219BK
Encompass_Preferred
MSH2 is involved in the DNA mismatch repair process. Mutations in the MSH2 gene contribute to the development of sporadic colorectal carcinoma. Immunostaining for MSH2 may be useful to identify the most probable gene responsible for MSI.
Catalog No. 50-826-06