Learn More
Aviva Systems Biology Corp MSH2 Antibody - N-terminal region (ARP41350_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41350_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MSH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MSH2. Alternate names: FCC1, COCA1, HNPCC, LCFS2, HNPCC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57815