Learn More
Cayman Chemical MSH3NTerm RabbIt Monclon 100uL
SDP
Catalog No. 502947075
Encompass_Preferred
A colorimetric reagent for the detection of iron forms a complex with ferrous iron that can be quantified by colorimetric detection at 562 nm as a measure of iron concentration
Catalog No. 50-294-7075 Supplier Cayman Chemical Supplier No. 32331100 UL
A colorimetric reagent for the detection of iron forms a complex with ferrous iron that can be quantified by colorimetric detection at 562 nm as a measure of iron concentration