Learn More
Aviva Systems Biology Corp MSH5 Antibody - N-terminal region (ARP41356_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41356_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSH5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MSH5. Alternate names: DKFZp434C1615, G7, MGC2939, MutSH5, NG23, MUTSH5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57819