Learn More
Biocare Medical, LLC MSH6 Antibody - Concentrate, 0.5 ml
SDP
Supplier:  Biocare Medical, LLC CM265BK
Encompass_Preferred
MSH6 is a heterodimer of MSH2 and binds to DNA containing G/T mismatches. MLH-1 and MSH2 are involved in the DNA mismatch repair (MMR) process. Microsatellite instability (MSI) is an alteration of microsatellite repeats during DNA replication and is a hallmark of the inactivation of the MMR genes. These defects in MMR have been related to human carcinogenesis. Studies have shown the mutations in MSH-1, MSH2 and MSH6 genes contribute to the development of sporadic colorectal carcinoma. MSH6 antibody may be useful to aid in identifying the most probable gene responsible for the MSI.
Catalog No. 50-826-27