Learn More
Medchemexpress LLC MSI-1436 lactate (trodusquemine lactate) | 1309370-86-2 | 98.8% | 100 MG
SDP
Catalog No. 5000230573
Encompass_Preferred
MSI-1436 lactate (trodusquemine lactate) is a selective, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) with reported IC50 ≈ 1 μM and approximately 200-fold selectivity over TCPTP. Supplied as a white to off-white solid, it is intended for biochemical and in vitro research applications.
  • Selective non-competitive PTP1B inhibitor (IC50 ≈ 1 μM).
  • Approximately 200-fold selectivity over TCPTP.
  • High purity suitable for research (98.8%).
  • Lactate salt form with molecular weight ~1000.17 g/mol.
  • Soluble in DMSO (~54 mg/mL) and 0.1 M HCl (~50 mg/mL) with warming and sonication.
  • Store solid at 4°C, sealed and away from moisture; store solutions at -20°C to -80°C.
Catalog No. 50-002-30573 Supplier Medchemexpress LLC Supplier No. HY12219A100MG
Add to Cart
Add to Cart
Request a Quote
MSI-1436 lactate (trodusquemine lactate) is a selective, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) with reported IC50 ≈ 1 μM and approximately 200-fold selectivity over TCPTP. Supplied as a white to off-white solid, it is intended for biochemical and in vitro research applications.
  • Selective non-competitive PTP1B inhibitor (IC50 ≈ 1 μM).
  • Approximately 200-fold selectivity over TCPTP.
  • High purity suitable for research (98.8%).
  • Lactate salt form with molecular weight ~1000.17 g/mol.
  • Soluble in DMSO (~54 mg/mL) and 0.1 M HCl (~50 mg/mL) with warming and sonication.
  • Store solid at 4°C, sealed and away from moisture; store solutions at -20°C to -80°C.