Learn More
Aviva Systems Biology Corp MSI2 Antibody - N-terminal region (ARP41112_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41112_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MSI2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MSI2. Alternate names: MSI2HApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57728