Learn More
Enzo Life Sciences MSK-195 (5mg). CAS: 289902-82-5
SDP
Supplier:  Enzo Life Sciences ALX550396M005
Encompass_Preferred
Potent analgesic with EC50=0.96µg/kg in the acetic acid-induced writhing test. RTX binding affinity (Ki=603nM), agonism (calcium influx; EC50=240nM). Alternative name: N-[2-(3,4-Dimethylbenzyl)-3-(pivalyloxy)propyl]-2-[4-(2-aminoethoxy)-3-methoxyphenyl]acetamide. Purity: ≥98%. Appearance: White solid. Solubility: Soluble in methanol, 100% ethanol, DMSO or other organic solvents; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9551