Learn More
Eurofins Discoverx MSK2, active, 250 µg. (14-616M)
Supplier:  Eurofins Discoverx 14616M
Encompass
N-terminal 6xHis-tagged recombinant, human MSK2 amino acids 2-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9060