Learn More
Carnabio Usa Inc MSK2(RPS6KA4)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 011485
Encompass
MSK2(RPS6KA4), Full-length, Wild type, Amino Acid 1-772, NP_003933.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0526