Learn More
Aviva Systems Biology Corp MSL1 Antibody - middle region (ARP64394_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64394_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSL1. It was validated on Western Blot. . Alternate names: DKFZp686J17211, MGC141861, MSL-1, hMSL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-254