Learn More
Aviva Systems Biology Corp MSL3L1 Antibody - N-terminal region (ARP39783_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39783_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSL3L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MSL3L1. Alternate names: DKFZP586J1822, MSL3L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-57177