Learn More
Thomas Scientific Microscope Slides, Diamond White Glass, 25 x 75mm, Charged, 90ø Ground Edges, Green Frosted, 72/Box
SDP
Catalog No. 501460275
Encompass
Positive charged microscope slides Color frosted: 1 end, 1 side 25 x 75 x 1mm Ground edges 90° corners Pre-cleaned 72 slides per box, 20 boxes per Case
Catalog No. 50-146-0275 Supplier Thomas Scientific Supplier No. 1184X37
Positive charged microscope slides Color frosted: 1 end, 1 side 25 x 75 x 1mm Ground edges 90° corners Pre-cleaned 72 slides per box, 20 boxes per Case