Learn More
New England Biolabs, Inc. MslI – 2500 units
SDP
Catalog No. 50812265
Encompass
A restriction endonuclease that recognizes the sequence CAYNN_^NNRTG.
Catalog No. 50-812-265 Supplier New England Biolabs, Inc. Supplier No. R0571L
A restriction endonuclease that recognizes the sequence CAYNN_^NNRTG.