Learn More
New England Biolabs, Inc. MslI – 500 units
SDP
Catalog No. 50812266
Encompass
A restriction endonuclease that recognizes the sequence CAYNN_^NNRTG.
Catalog No. 50-812-266 Supplier New England Biolabs, Inc. Supplier No. R0571S
A restriction endonuclease that recognizes the sequence CAYNN_^NNRTG.