Learn More
Thomas Scientific Microscope Slides, White Glass, 25 x 75mm, Positive Charged, 45ø Beveled Edges, Safety Corners , White Frosted, 72/Box
SDP
Catalog No. 501460404
Encompass
Produced from the finest quality white glass, Globe Scientific's charged microscope slides provide the ideal surface for cell and tissue adhesion. These popular slides are ideal for use in histology, cytology and pathology departments, where the convenience of an adhesive slide saves the user time and money. They are available in assorted colors and are pre-cleaned and ready to use. Produced from the finest quality optical white glass Charged surface allows cells and tissues to adhere to the slide Packed in high-quality white plastic boxes Dimensions: 25 x 75mm Thickness: 1 - 1.2mm Packaging: 72/plastic box, 20 boxes/case (10/gross)
Catalog No. 50-146-0404 Supplier Thomas Scientific Supplier No. 1186N60
Produced from the finest quality white glass, Globe Scientific's charged microscope slides provide the ideal surface for cell and tissue adhesion. These popular slides are ideal for use in histology, cytology and pathology departments, where the convenience of an adhesive slide saves the user time and money. They are available in assorted colors and are pre-cleaned and ready to use. Produced from the finest quality optical white glass Charged surface allows cells and tissues to adhere to the slide Packed in high-quality white plastic boxes Dimensions: 25 x 75mm Thickness: 1 - 1.2mm Packaging: 72/plastic box, 20 boxes/case (10/gross)