Learn More
Thomas Scientific Microscope Slides, Diamond White Glass, 25 x 75mm, 90ø Ground Edges, BLUE Frosted, 72/Box, 20 Boxes/Case (10 Gross)
SDP
Supplier:  Thomas Scientific 1208T55
Encompass
Diamond™ White Glass color coded microscope slides are offered in bright, attractive colors. They are available with ground edges and 90º corners, or with beveled edges and clipped corners. The slides are pre-cleaned and ready to use. Produced from the finest quality optical white glass Color coded on one end, one side Packed in high-quality white plastic boxes Dimensions: 25 x 75mm Thickness: 1mm Packaging: 72/plastic box, 20 boxes/case (10/gross)
Catalog No. 50-146-0257