Learn More
Thomas Scientific Microscope Slides, Diamond White Glass, 25 x 75mm, 90ø Ground Edges, Frosted: 1 End, 1 Side, 72/Box, 20 Boxes/Case (10 Gross)
SDP
Catalog No. 501460266
Encompass_Preferred
High quality Optically clear Made from chemically pure white glass sheets, the low iron content of these slides results in superior transparency, very low self-fluorescence and no color distortion. The glass is specially formulated for minimal oxidation, which ensures sample integrity during long-term storage. Increased hardness and molecular stability of the slide surface guarantees more accurate experimental results and diagnoses. Available in plain, frosted and color frosted versions. All slides are double cellophane packed to protect against moisture and foreign particles.
Catalog No. 50-146-0266 Supplier Thomas Scientific Supplier No. 1208T52
High quality Optically clear Made from chemically pure white glass sheets, the low iron content of these slides results in superior transparency, very low self-fluorescence and no color distortion. The glass is specially formulated for minimal oxidation, which ensures sample integrity during long-term storage. Increased hardness and molecular stability of the slide surface guarantees more accurate experimental results and diagnoses. Available in plain, frosted and color frosted versions. All slides are double cellophane packed to protect against moisture and foreign particles.