Learn More
Thomas Scientific Microscope Slides, Glass, 25 x 75mm, 45ø Beveled Edges, Clipped Corners, White Frosted
SDP
Catalog No. 501460392
Encompass
Dimensions: 25 x 75 x 1mm Thickness: 1.1 mm Color Frosted one end, one side Transportation and storage conditions considerably affect the quality and performance of microscope slides. Microscope slides should be stored in a dry place at constant room temperature. Slides should be stored on a pallet or shelf and not directly on a floor. Do not store slides and solvents together as the slide surface can turn hydrophobic. If the temperature is constantly warming and cooling, condensation may occur, which can cause sticking of the slides. We also recommend, when shipping to a customer, instruct them to allow the slides to reach room temperature prior to opening the cartons and using them. Since soda lime glass is subject to an aging process, we further recommend continuously rotating stock so the oldest slides are always used first. This will ensure that all slides being used are of the highest quality. Standard shelf life for soda lime glass microscope slides is approximately 2 years if t
Catalog No. 50-146-0392 Supplier Thomas Scientific Supplier No. 1192M13
Dimensions: 25 x 75 x 1mm Thickness: 1.1 mm Color Frosted one end, one side Transportation and storage conditions considerably affect the quality and performance of microscope slides. Microscope slides should be stored in a dry place at constant room temperature. Slides should be stored on a pallet or shelf and not directly on a floor. Do not store slides and solvents together as the slide surface can turn hydrophobic. If the temperature is constantly warming and cooling, condensation may occur, which can cause sticking of the slides. We also recommend, when shipping to a customer, instruct them to allow the slides to reach room temperature prior to opening the cartons and using them. Since soda lime glass is subject to an aging process, we further recommend continuously rotating stock so the oldest slides are always used first. This will ensure that all slides being used are of the highest quality. Standard shelf life for soda lime glass microscope slides is approximately 2 years if t