Learn More
Aviva Systems Biology Corp MSLN Antibody - middle region (ARP42262_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42262_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSLN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MSLN. Alternate names: CAK1, MPF, SMR, SMRPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58165