Learn More
Genscript Corporation MSLN/Mesothelin298-600 His
Supplier:  Genscript Corporation Z03876500
Encompass
Mesothelin also known as MSLN is a protein that in humans is encoded by the MSLN gene Cloning studies showed that the mesothelin gene encodes a precursor protein that is processed to yield mesothelin which is attached to the cell membrane by a glycophosphatidylinositol linkage and a 31-kDa shed fragment named megakaryocyte-potentiating factor (MPF)
Catalog No. 50-000-59448