Learn More
Aviva Systems Biology Corp MSMB Antibody - C-terminal region (ARP64207_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64207_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSMB. It was validated on Western Blot. . Alternate names: HPC13, IGBF, MSP, MSPB, PN44, PRPS, PSP, PSP-94, PSP57, PSP94ApplicationWB. Species reactivity: Human
Catalog No. 50-288-205