Learn More
Hoefer, Inc Mighty Small precast gels, 10% Bis-Tris, 12wells
SDP
Catalog No. NC3655161
Encompass_Preferred
"Mighty Small precast gels Hoefer Mighty Small precast gels are high-performance mini polyacrylamide gels commonly used for protein separation. Utilizing the Bis-Tris buffer system, they effectively minimize protein modifications during electrophoresis, resulting in sharper and clearer protein bands with higher resolution. High Loading Capacity Maximum sample volume is 50 μl Good Repeatability Fully automated gel casting process, minimal batch-to-batch variation, high stability, and good repeatability High Resolution Upgraded Bis-Trissystem for clearer, sharper separation bands and higher resolution Highly Effective Maximum 40 min for electrophoresis Ready to use and easy to store No solution preparation or gel casting is required, saving time. Store at 2-8°C Safe and non-toxic No hazardous reagents need to be handled Specifications Plate Size: 10×8 cm Wells: 12-well, 15-well Concentration: • Gradient gels: 4-12%
Catalog No. NC3655161 Supplier Hoefer, Inc Supplier No. MSG010T
"Mighty Small precast gels Hoefer Mighty Small precast gels are high-performance mini polyacrylamide gels commonly used for protein separation. Utilizing the Bis-Tris buffer system, they effectively minimize protein modifications during electrophoresis, resulting in sharper and clearer protein bands with higher resolution. High Loading Capacity Maximum sample volume is 50 μl Good Repeatability Fully automated gel casting process, minimal batch-to-batch variation, high stability, and good repeatability High Resolution Upgraded Bis-Trissystem for clearer, sharper separation bands and higher resolution Highly Effective Maximum 40 min for electrophoresis Ready to use and easy to store No solution preparation or gel casting is required, saving time. Store at 2-8°C Safe and non-toxic No hazardous reagents need to be handled Specifications Plate Size: 10×8 cm Wells: 12-well, 15-well Concentration: • Gradient gels: 4-12%