Learn More
Aviva Systems Biology Corp Msn Antibody - C-terminal region (ARP56189_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56189_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Msn. It was validated on Western Blot. . Alternate names: C78546ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-125