Learn More
Abcam MSOP, group III metabotropic glutamate receptor antagonist, 10MG
Catalog No. 502653051
Encompass_Preferred
MW 199.1 Da. Potent, selective group III metabotropic glutamate receptor antagonist. Selective inhibition for L-AP4-sensitive presynaptic mGluR on primary afferent terminals in spinal cord (KD = 51 μM) compared to the (1S,3S)-ACPD-sensitive presynaptic mGlu (KD > 700 μM). No activity on postsynaptic mGlu receptors or ionotropic glutamate receptors on neonatal rat motoneurons. Also available in simple stock solutions (ab146735) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-3051 Supplier Abcam Supplier No. AB14138110MG
MW 199.1 Da. Potent, selective group III metabotropic glutamate receptor antagonist. Selective inhibition for L-AP4-sensitive presynaptic mGluR on primary afferent terminals in spinal cord (KD = 51 μM) compared to the (1S,3S)-ACPD-sensitive presynaptic mGlu (KD > 700 μM). No activity on postsynaptic mGlu receptors or ionotropic glutamate receptors on neonatal rat motoneurons. Also available in simple stock solutions (ab146735) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement