Learn More
New England Biolabs, Inc. MspI Methyltransferase – 100 units
SDP
Catalog No. 50811634
Encompass
MspI Methyltransferase recognizes the same sequence as the HpaII Methyltransferase, but modifies the external cytosine residue (C5) of the sequence CCGG.
Catalog No. 50-811-634 Supplier New England Biolabs, Inc. Supplier No. M0215S
MspI Methyltransferase recognizes the same sequence as the HpaII Methyltransferase, but modifies the external cytosine residue (C5) of the sequence CCGG.