Learn More
New England Biolabs, Inc. MspA1I – 2500 units
SDP
Catalog No. 50812277
Encompass
A restriction endonuclease that recognizes the sequence CMG_^CKG.
Catalog No. 50-812-277 Supplier New England Biolabs, Inc. Supplier No. R0577L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
A restriction endonuclease that recognizes the sequence CMG_^CKG.