Learn More
New England Biolabs, Inc. MspA1I – 500 units
SDP
Catalog No. 50812278
Encompass
A restriction endonuclease that recognizes the sequence CMG_^CKG.
Catalog No. 50-812-278 Supplier New England Biolabs, Inc. Supplier No. R0577S
A restriction endonuclease that recognizes the sequence CMG_^CKG.