Learn More
Biotage MSPD15ML SGLFRTRS 100/PK 100
SDP
Catalog No. 502311417
Encompass
Reservoir, Single fritted; Biotage; Isolute; MSPD; For SPE; Volume: 15mL; Fitted with a single bottom frit (D); 100 additional frits included; 100/pk.
Catalog No. 50-231-1417 Supplier Biotage Supplier No. 1201143D
Reservoir, Single fritted; Biotage; Isolute; MSPD; For SPE; Volume: 15mL; Fitted with a single bottom frit (D); 100 additional frits included; 100/pk.