Learn More
New England Biolabs, Inc. MspI – 25000 units
SDP
Catalog No. 50811932
Encompass
A restriction endonuclease that recognizes the sequence C^CG_G.
Catalog No. 50-811-932 Supplier New England Biolabs, Inc. Supplier No. R0106L
A restriction endonuclease that recognizes the sequence C^CG_G.