Learn More
New England Biolabs, Inc. MspI – 5000 units
SDP
Catalog No. 50811934
Encompass
A restriction endonuclease that recognizes the sequence C^CG_G.
Catalog No. 50-811-934 Supplier New England Biolabs, Inc. Supplier No. R0106S
A restriction endonuclease that recognizes the sequence C^CG_G.