Learn More
New England Biolabs, Inc. MspJI – 1000 units
SDP
Catalog No. 504463
Encompass
A restriction endonuclease that recognizes the sequence CNNRNNNNNNNNN^NNNN_.
Catalog No. 50-446-3 Supplier New England Biolabs, Inc. Supplier No. R0661L
A restriction endonuclease that recognizes the sequence CNNRNNNNNNNNN^NNNN_.