Learn More
New England Biolabs, Inc. MspJI – 200 units
SDP
Catalog No. 501047489
Encompass
A restriction endonuclease that recognizes the sequence CNNRNNNNNNNNN^NNNN_.
Catalog No. 50-104-7489 Supplier New England Biolabs, Inc. Supplier No. R0661S
A restriction endonuclease that recognizes the sequence CNNRNNNNNNNNN^NNNN_.