Learn More
Analytical Sales & Services MSQ Vial Kit: Amber Screw Vials and Caps with PTFE/Silicone Liners
Catalog No. 5000805004
Encompass
Advantage™ Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Screw Vials and 9mm Screw Caps with MSQ PTFE/Silicone Liners
Catalog No. 50-008-05004 Supplier Analytical Sales & Services Supplier No. 99005ACASE
Advantage™ Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Screw Vials and 9mm Screw Caps with MSQ PTFE/Silicone Liners