Learn More
Analytical Sales & Services MSQ Vial Kit: Amber Screw Vials w/ 300µL Fused Insert and Marking Spot, Caps with PTFE/Silicone Liners
Catalog No. 5000805663
Encompass
Advantage™ Unassembled Custom Vial Kit. Contains: 9mm, 2mL Amber Glass Screw Vials w/ 300µL Fused Glass Insert and Marking Spot, 9mm Screw Caps with MSQ PTFE/Silicone Liners
Catalog No. 50-008-05663 Supplier Analytical Sales & Services Supplier No. 99003AM
Advantage™ Unassembled Custom Vial Kit. Contains: 9mm, 2mL Amber Glass Screw Vials w/ 300µL Fused Glass Insert and Marking Spot, 9mm Screw Caps with MSQ PTFE/Silicone Liners