Learn More
Analytical Sales & Services MSQ Vial Kit: Amber Snap/Crimp Vials with Marking Spot and Caps with Pre-Slit PTFE/Silicone Liners
Catalog No. 5000805928
Encompass
Advantage™ Unassembled Precision MSQ Snap Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Snap/Crimp Vials with Marking Spots and 9mm Snap Caps with Pre-Slit MSQ PTFE/Silicone Liners
Catalog No. 50-008-05928 Supplier Analytical Sales & Services Supplier No. 99015AMSLCASE
Advantage™ Unassembled Precision MSQ Snap Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Snap/Crimp Vials with Marking Spots and 9mm Snap Caps with Pre-Slit MSQ PTFE/Silicone Liners