Learn More
Analytical Sales & Services MSQ Vial Kit: Amber Snap/Crimp Vials with Marking Spot and Caps with PTFE/Silicone Liners
Catalog No. 5000805870
Encompass
Advantage™ Unassembled Precision MSQ Snap Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Snap/Crimp Vials with Marking Spots and 9mm Snap Caps with MSQ PTFE/Silicone Liners
Catalog No. 50-008-05870 Supplier Analytical Sales & Services Supplier No. 99015AMCASE
Advantage™ Unassembled Precision MSQ Snap Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Snap/Crimp Vials with Marking Spots and 9mm Snap Caps with MSQ PTFE/Silicone Liners