Learn More
Analytical Sales & Services MSQ VIAL KIT CLEAR SCREW 300UL
Catalog No. NC2245789
Encompass
MSQ Vial Kit: Clear Screw Vials w/ 300µL Fused Glass Insert and Marking Spot, Caps with Pre-Slit PTFE/Silicone Liner
This item is not returnable. View return policy
Catalog No. NC2245789 Supplier Analytical Sales & Services Supplier No. 99003MSL
MSQ Vial Kit: Clear Screw Vials w/ 300µL Fused Glass Insert and Marking Spot, Caps with Pre-Slit PTFE/Silicone Liner