Learn More
Analytical Sales & Services MSQ Vial Kit: Clear Screw Vials and Caps with PTFE/Silicone Liners
Catalog No. NC1288163
Encompass
Advantage Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Screw Vials and 9mm Screw Caps with MSQ PTFE/Silicone Liners
Catalog No. NC1288163 Supplier Analytical Sales & Services Supplier No. 99005CASE
Advantage Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Screw Vials and 9mm Screw Caps with MSQ PTFE/Silicone Liners