Learn More
Analytical Sales & Services MSQ Vial Kit: Clear Snap/Crimp Vials with Marking Spot and Caps with PTFE/Silicone Liners
Catalog No. 5000805929
Encompass
Advantage™ Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Snap/Crimp Vials with Marking Spots and 9mm Snap Caps with MSQ PTFE/Silicone Liners
Catalog No. 50-008-05929 Supplier Analytical Sales & Services Supplier No. 99015MCASE
Advantage™ Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Snap/Crimp Vials with Marking Spots and 9mm Snap Caps with MSQ PTFE/Silicone Liners