Learn More
Analytical Sales & Services MSQ Vial Kit: ClearScrew Vials w/ 300µL Fused Glass Insert and Marking Spot, Caps with PTFE/Silicone Liners
Catalog No. 5000805831
Encompass
Advantage™ Unassembled Custom Vial Kit. Contains: 9mm, 2mL Glass Screw Vials w/ 300µL Fused Glass Insert and Marking Spot, 9mm Screw Cap with MSQ PTFE/Silicone Liners
Catalog No. 50-008-05831 Supplier Analytical Sales & Services Supplier No. 99003M
Advantage™ Unassembled Custom Vial Kit. Contains: 9mm, 2mL Glass Screw Vials w/ 300µL Fused Glass Insert and Marking Spot, 9mm Screw Cap with MSQ PTFE/Silicone Liners