Learn More
CAS DATALOGGERS MSR 175 Plus Shock/Vibration, Temperature, Humidity, Pressure, and Light Transit Data Logger with GPS
Catalog No. NC3847104
Encompass
The MSR175 V1 is a small, robust, stand-alone data logger for transportation applications. It utilizes high-resolution sensors to continuously record what happens to your goods during transit. The data can be used to ascertain possible damage or used to improve transport and shipping packaging. The logger features two internal 3-axis accelerometers that can record +/- 15g and +/-100g as well as an internal temperature (-20C to 65C) and humidity sensor.
Catalog No. NC3847104 Supplier CAS DATALOGGERS Supplier No. MSR175PLUSC
The MSR175 V1 is a small, robust, stand-alone data logger for transportation applications. It utilizes high-resolution sensors to continuously record what happens to your goods during transit. The data can be used to ascertain possible damage or used to improve transport and shipping packaging. The logger features two internal 3-axis accelerometers that can record +/- 15g and +/-100g as well as an internal temperature (-20C to 65C) and humidity sensor.