Learn More
CAS DATALOGGERS MSR 175 Plus Shock/Vibration, Temperature, and Humidity Transit Data Logger
Catalog No. NC3847105
Encompass
"The MSR175 V1 is a small, robust, stand-alone data logger for transportation applications and is air cargo approved. It utilizes high-resolution sensors to continuously record what happens to your goods during transit. The data can be used to ascertain possible damage or used to improve transport and shipping packaging. The logger features two internal 3-axis accelerometers that can record +/- 15g and +/-100g as well as an internal temperature (-20C to 65C) and humidity sensor.The internal non-volatile memory is capable of storing over 2 million measurement values which is enough for up to 1000 shock events. The MSR175 is built with an internal 230 mAh LiPo-battery which when fully charged can power the logger for a recording period of up to 8 weeks."
Catalog No. NC3847105 Supplier CAS DATALOGGERS Supplier No. MSR175V1
"The MSR175 V1 is a small, robust, stand-alone data logger for transportation applications and is air cargo approved. It utilizes high-resolution sensors to continuously record what happens to your goods during transit. The data can be used to ascertain possible damage or used to improve transport and shipping packaging. The logger features two internal 3-axis accelerometers that can record +/- 15g and +/-100g as well as an internal temperature (-20C to 65C) and humidity sensor.The internal non-volatile memory is capable of storing over 2 million measurement values which is enough for up to 1000 shock events. The MSR175 is built with an internal 230 mAh LiPo-battery which when fully charged can power the logger for a recording period of up to 8 weeks."