Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Msr-blue | 2966537-39-1 | 99.4% | 222.26 | C11H10O3S | 10 MG
SDP

Catalog No. 5000296981
Encompass_Preferred

Msr-blue is a turn-on fluorescent probe for detecting methionine sulfoxide reductase activity in vitro and in live cells. It yields a large fluorescence increase upon enzymatic activation and is provided as a solid for preparation of DMSO stock solutions.

  • High sensitivity: more than 100-fold fluorescence increase when activated by methionine sulfoxide reductase.
  • Suitable for live-cell assays: compatible with imaging at λex 340 nm and λem 440 nm.
  • High purity: typical purity ~99.4%.
  • Solubility: soluble in DMSO at 25 mg/mL; may require ultrasonication.
  • Storage: store solid protected from light at 4°C; prepared solutions stable at -80°C for long-term storage.

Catalog No. 50-002-96981 Supplier Medchemexpress LLC Supplier No. HYD125610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Msr-blue is a turn-on fluorescent probe for detecting methionine sulfoxide reductase activity in vitro and in live cells. It yields a large fluorescence increase upon enzymatic activation and is provided as a solid for preparation of DMSO stock solutions.

  • High sensitivity: more than 100-fold fluorescence increase when activated by methionine sulfoxide reductase.
  • Suitable for live-cell assays: compatible with imaging at λex 340 nm and λem 440 nm.
  • High purity: typical purity ~99.4%.
  • Solubility: soluble in DMSO at 25 mg/mL; may require ultrasonication.
  • Storage: store solid protected from light at 4°C; prepared solutions stable at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.