Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Msr-ratio (Msr-green) | 2290635-22-0 | 99.1% | 310.37 | C18H14O3S | 10 MG
SDP

Catalog No. 5000296784
Encompass_Preferred

Msr-Ratio (Msr-green) is a ratiometric fluorescent probe for monitoring methionine sulfoxide reductase (Msr) activity in vitro and in live cells. It exhibits excitation at 375 nm and emission at 550 nm, and is supplied at high purity for research applications.

  • Ratiometric fluorescent probe for methionine sulfoxide reductase activity.
  • Excitation 375 nm and emission 550 nm for sensitive detection.
  • High purity (99.13%) suitable for cell-based and in vitro assays.
  • Molecular weight 310.37 for compact chemical profile.
  • Available in small quantities such as 10 mg for research workflows.

Catalog No. 50-002-96784 Supplier Medchemexpress LLC Supplier No. HYD125710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Msr-Ratio (Msr-green) is a ratiometric fluorescent probe for monitoring methionine sulfoxide reductase (Msr) activity in vitro and in live cells. It exhibits excitation at 375 nm and emission at 550 nm, and is supplied at high purity for research applications.

  • Ratiometric fluorescent probe for methionine sulfoxide reductase activity.
  • Excitation 375 nm and emission 550 nm for sensitive detection.
  • High purity (99.13%) suitable for cell-based and in vitro assays.
  • Molecular weight 310.37 for compact chemical profile.
  • Available in small quantities such as 10 mg for research workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.