Learn More
Cayman Chemical MSR1CD204 RabbIt MoncLNal 50uL
SDP
Catalog No. 502916554
Encompass_Preferred
A Notch1 signaling modulator decreases the production of NB25 from full-length Notch1 in HEK293 cells decreases activation of HES1 in a luciferase reporter assay using HCT116 cells
Catalog No. 50-291-6554 Supplier Cayman Chemical Supplier No. 3809050 UL
A Notch1 signaling modulator decreases the production of NB25 from full-length Notch1 in HEK293 cells decreases activation of HES1 in a luciferase reporter assay using HCT116 cells