Learn More
NEOGEN Modified Semi-Solid Rappaport-Vassiliadis (MSRV) Agar, 10 kg
SDP
Catalog No. 502015350
Encompass_Preferred
Modified Semi-Solid Rappaport-Vassiliadis (MSRV) Agar is used with Novobiocin for the rapid detection of motile Salmonella spp. and is not intended for use in the diagnosis of disease or other conditions in humans. MSRV was developed in 1986 by De Smedt, Bolderdijk and Rappold as a rapid means of Salmonella detection. The medium, based upon Rappaport Vassiliadis broth, is inoculated directly from the preenrichment medium, in the center of the plate. Motile organisms spread from the center in the semi-solid agar, but non-salmonellas are inhibited by the selective agents. After overnight incubation the use of polyvalent salmonella antisera or a latex kit can confirm the presence of a Salmonella.For more information visit https://www.neogen.com/solutions/Microbiology/modified-semi-solid-rappaport-vassiliadis-agar
Catalog No. 50-201-5350 Supplier NEOGEN Supplier No. 700003163
Modified Semi-Solid Rappaport-Vassiliadis (MSRV) Agar is used with Novobiocin for the rapid detection of motile Salmonella spp. and is not intended for use in the diagnosis of disease or other conditions in humans. MSRV was developed in 1986 by De Smedt, Bolderdijk and Rappold as a rapid means of Salmonella detection. The medium, based upon Rappaport Vassiliadis broth, is inoculated directly from the preenrichment medium, in the center of the plate. Motile organisms spread from the center in the semi-solid agar, but non-salmonellas are inhibited by the selective agents. After overnight incubation the use of polyvalent salmonella antisera or a latex kit can confirm the presence of a Salmonella.For more information visit https://www.neogen.com/solutions/Microbiology/modified-semi-solid-rappaport-vassiliadis-agar