Learn More
TARGETMOL CHEMICALS INC MST-312 25MG
SDP
Catalog No. 502579939
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MST-312 (Telomerase Inhibitor IX) is a derivative of green tea epigallocatechin gallate (EGCG). MST-312 is a telomerase inhibitor. MST-312 has research value in cancer such as multiple myeloma (MM). purity: 99%
Catalog No. 50-257-9939 Supplier TARGETMOL CHEMICALS INC Supplier No. T4129025MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MST-312 (Telomerase Inhibitor IX) is a derivative of green tea epigallocatechin gallate (EGCG). MST-312 is a telomerase inhibitor. MST-312 has research value in cancer such as multiple myeloma (MM). purity: 99%