Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MST-312, chemically modified derivative of epigallocatechin gallate | 368449-04-1 | 98.6% | 100 MG
SDP

Catalog No. 5000232585
Encompass_Preferred

MST-312 is a small-molecule telomerase inhibitor derived from epigallocatechin gallate (EGCG), intended for research use in oncology and cellular studies. It is available both as a solid and as a DMSO solution for in vitro assays.

  • Telomerase inhibitor used in cancer research.
  • Purity 98.6%.
  • Molecular formula C20H16N2O6.
  • Molecular weight 380.35 g/mol.
  • CAS number 368449-04-1.
  • Available as solid (5-500 mg) and 10 mM solution in DMSO.

Catalog No. 50-002-32585 Supplier Medchemexpress LLC Supplier No. HY120145100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MST-312 is a small-molecule telomerase inhibitor derived from epigallocatechin gallate (EGCG), intended for research use in oncology and cellular studies. It is available both as a solid and as a DMSO solution for in vitro assays.

  • Telomerase inhibitor used in cancer research.
  • Purity 98.6%.
  • Molecular formula C20H16N2O6.
  • Molecular weight 380.35 g/mol.
  • CAS number 368449-04-1.
  • Available as solid (5-500 mg) and 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.